This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other WSB2 SIRNA Assays ›
Gene Symbol:
Wsb2
Gene Name:
WD repeat and SOCS box-containing 2
Gene Aliases:
AA673511, Swip2, WSB-2
Chromosome Location:
Chr.5: 117357305 - 117378589 on Build GRCm38
UniGene ID:
Mm.28489
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 162741

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Wsb2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021539.4 NP_067514.2 9 1879
GenBank mRNA AK150710.1 - 7 1453
AK195234.1 - 1 57
AK198504.1 - 1 57
AK206996.1 - 1 57
BC055100.1 - 9 1879 6589250

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
intracellular signal transduction
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mlg7h:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline