This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CTDP1 SIRNA Assays ›
Gene Symbol:
Ctdp1
Gene Name:
CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) phosphatase, subunit 1
Gene Aliases:
4930563P03Rik, AW553592
Chromosome Location:
Chr.18: 80407959 - 80470612 on Build GRCm38
UniGene ID:
Mm.312893
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 163027

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ctdp1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026295.2 NP_080571.2 4 704
GenBank mRNA AK156617.1 - 4 733
BC053435.1 - 4 704 6394578

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001106131, XM_006254942, XM_006254943, XM_006254944.

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
cell cycle
mitotic nuclear division
exit from mitosis
cell division
negative regulation of cell growth involved in cardiac muscle cell development
phosphoprotein phosphatase activity
protein binding
CTD phosphatase activity
hydrolase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline