siRNA Details
| Gene Symbol: Tshz3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172298.2 | NP_758502.1 | 2 | 959 | ||
| GenBank mRNA | AK220396.1 | - | 2 | 830 | ||
| AY063491.1 | - | 2 | 959 | |||
| BC150674.1 | - | 2 | 902 | 9087585 | ||
UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Gene Symbol: Tshz3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172298.2 | NP_758502.1 | 2 | 959 | ||
| GenBank mRNA | AK220396.1 | - | 2 | 830 | ||
| AY063491.1 | - | 2 | 959 | |||
| BC150674.1 | - | 2 | 902 | 9087585 | ||
Tshz3
Genomic Map