This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LAMA5 SIRNA Assays ›
Gene Symbol:
LAMA5
Gene Name:
laminin subunit alpha 5
Gene Aliases:
-
Chromosome Location:
Chr.20: 62309060 - 62367318 on Build GRCh38
UniGene ID:
Hs.473256
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 16425

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LAMA5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005560.4 NP_005551.3 12 1642
XM_006723796.3 XP_006723859.1 12 1645
XM_006723798.3 XP_006723861.1 12 1648
XM_011528818.2 XP_011527120.1 12 1645
XM_011528819.2 XP_011527121.1 12 1645
GenBank mRNA AB067494.1 - 9 986
AF443072.1 - 12 1642

Back To Top

More Information


Panther Classification:

Molecular Function -

extracellular matrix protein

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
branching involved in ureteric bud morphogenesis
morphogenesis of a polarized epithelium
neural crest cell migration
hair follicle development
cytoskeleton organization
integrin-mediated signaling pathway
muscle organ development
cell recognition
cell proliferation
embryo development
morphogenesis of embryonic epithelium
cell migration
cell differentiation
regulation of cell adhesion
extracellular matrix organization
lung development
regulation of cell migration
substrate adhesion-dependent cell spreading
regulation of cell proliferation
cilium assembly
odontogenesis of dentin-containing tooth
endothelial cell differentiation
regulation of embryonic development
focal adhesion assembly
branching involved in salivary gland morphogenesis
establishment of protein localization to plasma membrane
integrin binding
structural molecule activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-9hvhj:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline