siRNA Details
| Gene Symbol: Garnl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178888.4 | NP_849219.2 | 21 | 1824 | ||
| GenBank mRNA | AK034068.1 | - | 21 | 1826 | ||
| AK036258.1 | - | 20 | 2122 | |||
| AK083673.1 | - | 5 | 396 | |||
| AK083773.1 | - | 2 | 1548 | |||
| AK133163.1 | - | 22 | 1998 | |||
| BC132143.1 | - | 21 | 1767 | 40130420 | ||

Garnl3
Genomic Map