This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SNCAIP SIRNA Assays ›
Gene Symbol:
SNCAIP
Gene Name:
synuclein alpha interacting protein
Gene Aliases:
SYPH1, Sph1
Chromosome Location:
Chr.5: 122311354 - 122464219 on Build GRCh38
UniGene ID:
Hs.426463
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 16449

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SNCAIP
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242935.2 NP_001229864.1 8 1096
NM_001308100.1 NP_001295029.1 12 3593
NM_001308105.1 NP_001295034.1 8 1752
NM_001308106.1 NP_001295035.1 8 1021
NM_001308107.1 NP_001295036.1 8 1096
NM_001308108.1 NP_001295037.1 8 1060
NM_001308109.1 NP_001295038.1 7 982
NM_005460.3 NP_005451.2 10 2148
NR_131761.1 12 2298
NR_131762.1 9 1027
XM_005272138.3 XP_005272195.1 10 2071
XM_005272139.1 XP_005272196.1 10 2011
XM_006714734.2 XP_006714797.1 10 2204
XM_011543737.2 XP_011542039.1 11 2206
XM_011543738.2 XP_011542040.1 11 2205
XM_011543739.1 XP_011542041.1 11 2131
XM_011543741.2 XP_011542043.1 11 2155
XM_011543743.2 XP_011542045.1 11 2580
XM_011543745.1 XP_011542047.1 10 2035
XM_011543749.2 XP_011542051.1 8 1386
XM_011543750.1 XP_011542052.1 7 864
XM_017010078.1 XP_016865567.1 11 4533
XM_017010079.1 XP_016865568.1 11 2206
XM_017010080.1 XP_016865569.1 11 2206
XM_017010081.1 XP_016865570.1 11 2204
XM_017010082.1 XP_016865571.1 10 2035
XM_017010083.1 XP_016865572.1 8 1012
XM_017010085.1 XP_016865574.1 9 1091
GenBank mRNA AB110788.1 - 9 1886
AB110789.1 - 8 1706
AB110790.1 - 12 2252
AB110791.1 - 8 975
AB110792.1 - 7 834
AB110793.1 - 8 1014
AB110794.1 - 6 720
AF076929.1 - 10 1979
AK001617.1 - 6 832
AK021944.1 - 3 871
AK298882.1 - 8 977
AK299687.1 - 9 1027
AK304646.1 - 7 814
BC033743.1 - 8 916 5193089
BC040552.1 - 12 3455 5298588
BC094759.1 - 8 974 4838145
DQ227317.1 - 8 1040

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell death
dopamine metabolic process
cellular protein metabolic process
regulation of neurotransmitter secretion
regulation of inclusion body assembly
protein binding
ubiquitin protein ligase binding
identical protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0