This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TBC1D31 SIRNA Assays ›
Gene Symbol:
Tbc1d31
Gene Name:
TBC1 domain family, member 31
Gene Aliases:
4B3, D330013L20Rik, Gm85, Wdr67
Chromosome Location:
Chr.15: 57911684 - 57970068 on Build GRCm38
UniGene ID:
Mm.390835
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 164539

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tbc1d31
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081396.2 NP_001074865.2 5 757
NM_001167679.1 NP_001161151.1 5 757
GenBank mRNA AB185844.1 - 5 725
AK084542.1 - 4 521
AK141805.1 - 5 627
AK154264.1 - 5 722
AK161403.1 - 4 574
AK171024.1 - 5 722
BC050039.1 - 5 703 30016266
BC066830.1 - 5 689 30463017

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline