This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SH2D3A SIRNA Assays ›
Gene Symbol:
SH2D3A
Gene Name:
SH2 domain containing 3A
Gene Aliases:
NSP1
Chromosome Location:
Chr.19: 6752160 - 6767513 on Build GRCh38
UniGene ID:
Hs.439645
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 16469

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SH2D3A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005490.2 NP_005481.2 5 997
XM_005259467.1 XP_005259524.1 5 947
XM_005259472.2 XP_005259529.1 4 560
XM_011527608.2 XP_011525910.1 4 1364
XM_011527609.2 XP_011525911.1 4 861
XM_017026123.1 XP_016881612.1 5 947
GenBank mRNA AF124249.1 - 5 954
AK225386.1 - 5 920
AK290293.1 - 5 1073
AK292781.1 - 5 920
AK299406.1 - 4 570
AK310659.1 - 5 920
AY358406.1 - 5 954
BC006281.2 - 5 934 3958342

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

JNK cascade
small GTPase mediated signal transduction
positive regulation of signal transduction
positive regulation of GTPase activity
SH3/SH2 adaptor activity
guanyl-nucleotide exchange factor activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-rdsp8:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline