siRNA Details
| Gene Symbol: Uimc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293660.1 | NP_001280589.1 | 10 | 981 | ||
| NM_011307.2 | NP_035437.1 | 12 | 1819 | |||
| GenBank mRNA | AK052704.1 | - | 12 | 1820 | ||
| AK083342.1 | - | 12 | 1760 | |||
| AK160926.1 | - | 10 | 982 | |||
| BC037092.1 | - | 11 | 1694 | |||
| BC040808.1 | - | 12 | 1775 | 4038778 | ||
| BC051659.1 | - | 12 | 1742 | |||
| BC066220.1 | - | 12 | 1742 | |||
| U22015.1 | - | 8 | 1271 | |||

Uimc1
Genomic Map