This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other KDM3B SIRNA Assays ›
Gene Symbol:
Kdm3b
Gene Name:
KDM3B lysine (K)-specific demethylase 3B
Gene Aliases:
5830462I21Rik, JHDM2B, Jmjd1b, mKIAA1082
Chromosome Location:
Chr.18: 34776243 - 34839370 on Build GRCm38
UniGene ID:
Mm.277906
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 166138

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kdm3b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081256.1 NP_001074725.1 14 3605
GenBank mRNA AK129281.1 - 13 2961
BC038376.1 - 4 423
BC060727.1 - 7 1165
BC108415.1 - 7 1009
BC151084.1 - 14 3566 9087997
BC172150.1 - 13 2966

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008772060.

Panther Classification:

Molecular Function -

histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
chromatin modification
oxidation-reduction process
response to cisplatin
sulfonate dioxygenase activity
molecular_function
antioxidant activity
oxidoreductase activity
2,4-dichlorophenoxyacetate alpha-ketoglutarate dioxygenase activity
procollagen-proline dioxygenase activity
hypophosphite dioxygenase activity
DNA-N1-methyladenine dioxygenase activity
metal ion binding
dioxygenase activity
C-19 gibberellin 2-beta-dioxygenase activity
C-20 gibberellin 2-beta-dioxygenase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nlplw:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline