This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other WDR37 SIRNA Assays ›
Gene Symbol:
Wdr37
Gene Name:
WD repeat domain 37
Gene Aliases:
-
Chromosome Location:
Chr.13: 8802966 - 8871904 on Build GRCm38
UniGene ID:
Mm.284654
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 166289

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Wdr37
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039388.2 NP_001034477.1 4, 5 633
NM_001039389.2 NP_001034478.1 4, 5 554
NM_172445.3 NP_766033.1 4, 5 554
GenBank mRNA AK014128.1 - 4, 5 530
AK032582.1 - 4, 5 516
AK036203.1 - 4, 5 550
AK044740.1 - 4, 5 534
AK129254.1 - 4, 5 510
AK146209.1 - 4, 5 529
BC046236.1 - 3, 4 381 6817089
BC048819.1 - 4, 5 478
CJ134699.1 - 4, 5 555

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-5lprg:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0