siRNA Details
| Gene Symbol: Fndc3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207636.2 | NP_997519.2 | 2, 3 | 432 | ||
| GenBank mRNA | AK032733.1 | - | 2, 3 | 404 | ||
| AK048586.1 | - | 2, 3 | 435 | |||
| AK089380.1 | - | 2, 3 | 395 | |||
| AK154756.1 | - | 2, 3 | 273 | |||
| AK154790.1 | - | 2, 3 | 202 | |||
| AK164398.1 | - | 2, 3 | 430 | |||
| AK172458.1 | - | 2, 3 | 296 | |||
| BC144880.1 | - | 2, 3 | 240 | |||
| DQ192036.1 | - | 2, 3 | 433 | |||

Fndc3a
Genomic Map