siRNA Details
| Gene Symbol: Rbak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045482.2 | NP_001038947.1 | 5 | 741 | ||
| NM_021326.3 | NP_067301.1 | 5 | 841 | |||
| GenBank mRNA | AF226870.1 | - | 5 | 699 | ||
| AK051009.1 | - | 5 | 842 | |||
| BC059913.1 | - | 3 | 600 | |||
| CN699599.1 | - | 5 | 535 | |||

Rbak
Genomic Map