siRNA Details
| Gene Symbol: Nlrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177690.3 | NP_808358.2 | 2 | 239 | ||
| GenBank mRNA | AK054264.1 | - | 2 | 240 | ||
| AY360473.1 | - | 2 | 240 | |||
| DQ066881.1 | - | 2 | 240 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nlrp2| Gene Symbol: Nlrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177690.3 | NP_808358.2 | 2 | 239 | ||
| GenBank mRNA | AK054264.1 | - | 2 | 240 | ||
| AY360473.1 | - | 2 | 240 | |||
| DQ066881.1 | - | 2 | 240 | |||