siRNA Details
| Gene Symbol: 1810024B03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198630.2 | NP_941032.1 | 2 | 871 | ||
| GenBank mRNA | AK052786.1 | - | 2 | 872 | ||
| BC117065.1 | - | 2 | 824 | 40126384 | ||
| BC119086.1 | - | 2 | 824 | 8733835 | ||

1810024B03Rik
Genomic Map