This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ATP6AP2 SIRNA Assays ›
Gene Symbol:
ATP6AP2
Gene Name:
ATPase H+ transporting accessory protein 2
Gene Aliases:
APT6M8-9, ATP6IP2, ATP6M8-9, ELDF10, HT028, M8-9, MRXE, MRXSH, MSTP009, PRR, RENR, XMRE, XPDS
Chromosome Location:
Chr.X: 40580964 - 40606637 on Build GRCh38
UniGene ID:
Hs.495960
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 16913

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ATP6AP2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005765.2 NP_005756.2 3 353
GenBank mRNA AF109363.1 - 3 314
AF248966.1 - 3 358
AF291814.1 - 3 340
AK075382.1 - 3 411
AK293510.1 - 3 335
AK296614.1 - 2 216
AK297007.1 - 3 324
AK297760.1 - 3 423
AK314633.1 - 3 304
AK315948.1 - 3 284
AL049929.1 - 2 193
AY038990.1 - 3 356
AY429341.1 - 2 216
BC084541.1 - 3 333 6575732

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-5d9f4b9bfc-8vt9q:80/100.66.74.193:80.
git-commit: 52aea788d112809596cd23e639b0bc7998ca5a01
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.40.1-2025.11.48-1.0