siRNA Details
| Gene Symbol: Slc39a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135151.1 | NP_001128623.1 | 10 | 3583 | ||
| NM_001135152.1 | NP_001128624.1 | 10 | 3549 | |||
| NM_144808.4 | NP_659057.2 | 10 | 3583 | |||
| GenBank mRNA | AB177995.1 | - | 11 | 2300 | ||
| AK122197.1 | - | 7 | 3046 | |||
| BC030883.1 | - | 1 | 1530 | |||

Slc39a14
Genomic Map