This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PRG4 SIRNA Assays ›
Gene Symbol:
PRG4
Gene Name:
proteoglycan 4
Gene Aliases:
CACP, HAPO, JCAP, MSF, SZP
Chromosome Location:
Chr.1: 186296273 - 186314562 on Build GRCh38
UniGene ID:
Hs.647723
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 16942

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PRG4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127708.2 NP_001121180.2 4 306
NM_001303232.1 NP_001290161.1 5 429
NM_005807.4 NP_005798.3 5 429
GenBank mRNA AK131434.1 - 3 290
AK301343.1 - 5 424
AK301487.1 - 3 300
DA977744.1 - 4 306
DA995681.1 - 5 429
U70136.1 - 4 411

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

receptor-mediated endocytosis
immune response
cell proliferation
scavenger receptor activity
polysaccharide binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline