siRNA Details
| Gene Symbol: Tmem184a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161548.1 | NP_001155020.1 | 2, 3 | 464 | ||
| NM_144914.3 | NP_659163.3 | 1, 2 | 284 | |||
| GenBank mRNA | AK050489.1 | - | 2, 3 | 434 | ||
| AK148454.1 | - | 256 | ||||
| BC026659.1 | - | 2, 3 | 423 | 4988857 | ||

Tmem184a
Genomic Map