siRNA Details
| Gene Symbol: Mfap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145426.2 | NP_663401.2 | 2 | 461 | ||
| NM_180599.1 | NP_850930.1 | 2 | 425 | |||
| GenBank mRNA | AK042988.1 | - | 2 | 409 | ||
| AK076071.1 | - | 2 | 427 | |||
| AK080405.1 | - | 2 | 405 | |||
| AK155122.1 | - | 2 | 439 | |||
| BC006828.1 | - | 2 | 420 | 3597878 | ||
| BC058567.1 | - | 1 | 369 | |||
| CF174749.1 | - | 2 | 425 | |||

Mfap3
Genomic Map