siRNA Details
| Gene Symbol: Tiprl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145513.4 | NP_663488.1 | 2 | 346 | ||
| GenBank mRNA | AK049119.1 | - | 2 | 294 | ||
| AK049784.1 | - | 2 | 332 | |||
| AK075774.1 | - | 2 | 326 | |||
| BC002098.1 | - | 2 | 291 | 3590157 | ||
| BC058250.1 | - | 2 | 285 | 6315314 | ||
| CF531065.1 | - | 2 | 348 | |||

Tiprl
Genomic Map