This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TIPRL SIRNA Assays ›
Gene Symbol:
Tiprl
Gene Name:
TIP41, TOR signalling pathway regulator-like (S. cerevisiae)
Gene Aliases:
1810011K17Rik
Chromosome Location:
Chr.1: 165212286 - 165282478 on Build GRCm38
UniGene ID:
Mm.21520
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 169856

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tiprl
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145513.4 NP_663488.1 7 2270
GenBank mRNA AK049784.1 - 7 2256

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

DNA damage checkpoint
negative regulation of protein phosphatase type 2A activity
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-5djdz:80/100.66.79.206:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline