This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DMRT2 SIRNA Assays ›
Gene Symbol:
Dmrt2
Gene Name:
doublesex and mab-3 related transcription factor 2
Gene Aliases:
Terra
Chromosome Location:
Chr.19: 25672391 - 25679013 on Build GRCm38
UniGene ID:
Mm.275572
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 170005

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dmrt2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145831.3 NP_665830.1 4 1463
GenBank mRNA AF080623.1 - 434
AF539811.1 - 4 1463
AK044891.1 - 4 1464
AK140337.1 - 4 1455
AK140347.1 - 4 1458
BC027669.1 - 1 412 1248080

Back To Top

More Information


Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of somitogenesis
positive regulation of transcription from RNA polymerase II promoter
embryonic skeletal system development
left/right pattern formation
positive regulation of myotome development
RNA polymerase II regulatory region sequence-specific DNA binding
core promoter proximal region sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein homodimerization activity
sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-75zlr:80/100.66.79.163:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline