This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PANK3 SIRNA Assays ›
Gene Symbol:
Pank3
Gene Name:
pantothenate kinase 3
Gene Aliases:
-
Chromosome Location:
Chr.11: 35769495 - 35791285 on Build GRCm38
UniGene ID:
Mm.255044
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 170006

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pank3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145962.2 NP_666074.1 2 505
GenBank mRNA AK039865.1 - 2 507
AK136305.1 - 2 195
BC027089.1 - 2 117 5355443
BC032188.1 - 2 551
CJ127650.1 - 2 408

Back To Top

More Information


Panther Classification:

Molecular Function -

kinase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

coenzyme A biosynthetic process
phosphorylation
nucleotide binding
pantothenate kinase activity
ATP binding
kinase activity
transferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-584499df9d-7vrs5:80/100.66.78.233:80.
git-commit: 256cec658627a60c8e1196038bf5905a13cd45ba
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0