siRNA Details
| Gene Symbol: Ctage5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165253.1 | NP_001158725.1 | 11 | 1010 | ||
| NM_001165254.1 | NP_001158726.1 | 11 | 1009 | |||
| NM_146034.3 | NP_666146.3 | 13 | 1108 | |||
| GenBank mRNA | AK029466.1 | - | 11 | 1009 | ||
| AK053010.1 | - | 12 | 1015 | |||
| AK087188.1 | - | 12 | 1023 | |||
| AK157358.1 | - | 11 | 1011 | |||
| BC018567.1 | - | 5 | 391 | |||
| BC024076.1 | - | 4 | 348 | |||
| BC026864.1 | - | 13 | 1066 | 2647214 | ||

Ctage5
Genomic Map