This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TMEM119 SIRNA Assays ›
Gene Symbol:
Tmem119
Gene Name:
transmembrane protein 119
Gene Aliases:
AW208946, BC025600, obif
Chromosome Location:
Chr.5: 113793729 - 113800358 on Build GRCm38
UniGene ID:
Mm.41681
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 170309

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem119
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146162.2 NP_666274.1 2 313
GenBank mRNA AK135223.1 - 344
AK170712.1 - 2 322
BC025600.1 - 2 318 5250899

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

ossification
osteoblast differentiation
cell differentiation
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-z8bsd:80/100.66.74.109:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0