siRNA Details
| Gene Symbol: Nhlrc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172501.2 | NP_766089.1 | 2 | 339 | ||
| GenBank mRNA | AK029465.1 | - | 2 | 334 | ||
| AK033167.1 | - | 2 | 339 | |||
| AK133986.1 | - | 289 | ||||
| AK162824.1 | - | 2 | 379 | |||
| AK220477.1 | - | 404 | ||||
| BC087903.1 | - | 2 | 326 | 30250042 | ||
| BC107356.1 | - | 2 | 327 | 40055875 | ||

Nhlrc3
Genomic Map