siRNA Details
| Gene Symbol: Vezt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031848.1 | - | 7 | 1070 | ||
| AK036017.1 | - | 7 | 1007 | |||
| AK041354.1 | - | 8 | 1173 | |||
| AK042540.1 | - | 7 | 1001 | |||
| AY753561.1 | - | 7 | 963 | |||
| AY753562.1 | - | 7 | 963 | |||
| BC056428.1 | - | 7 | 994 | 5716245 | ||

Vezt
Genomic Map