This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ADAMTSL1 SIRNA Assays ›
Gene Symbol:
Adamtsl1
Gene Name:
ADAMTS-like 1
Gene Aliases:
5930437A14Rik, 6720426B09Rik, ADAMTSL-1
Chromosome Location:
Chr.4: 85513234 - 86428390 on Build GRCm38
UniGene ID:
Mm.331690
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 170816

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Adamtsl1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029967.3 NP_084243.3 21 4190
GenBank mRNA AK031246.1 - 4 793
AK132420.1 - 9 2630
BC116295.1 - 4 784
BC116296.1 - 4 784

Back To Top

More Information


Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
peptidase activity
metallopeptidase activity
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-vc5lg:80/100.66.78.61:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0