This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TMEM194B SIRNA Assays ›
Gene Symbol:
Tmem194b
Gene Name:
transmembrane protein 194B
Gene Aliases:
5330401P04Rik, 9830163O08
Chromosome Location:
Chr.1: 52568535 - 52651919 on Build GRCm38
UniGene ID:
Mm.83541
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 170997

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem194b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142647.1 NP_001136119.1 1, 2 146
GenBank mRNA AK030361.1 - 1, 2 115
AK036693.1 - 1, 2 106
AK048484.1 - 1, 2 103
AK086801.1 - 1, 2 57
BB848894.1 - 1, 2 149
CD553439.1 - 1, 2 74

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline