This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FBXW8 SIRNA Assays ›
Gene Symbol:
Fbxw8
Gene Name:
F-box and WD-40 domain protein 8
Gene Aliases:
4930438M06Rik, FBW6, FBW8, FBXO29, Fbx29
Chromosome Location:
Chr.5: 118064981 - 118155458 on Build GRCm38
UniGene ID:
Mm.221769
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 171114

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fbxw8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172721.2 NP_766309.2 3 591
GenBank mRNA AK034447.1 - 3 519
AK047695.1 - 3 559
AK047756.1 - 3 533
AK053794.1 - 3 592
AK134291.1 - 3 598
AK219233.1 - 1 43

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107145, XM_008769233.

Gene Ontology Categories:

Function(s) Process(es)

ubiquitin-dependent protein catabolic process
Golgi organization
cell proliferation
protein ubiquitination
positive regulation of dendrite morphogenesis
spongiotrophoblast layer development
labyrinthine layer blood vessel development
ubiquitin-protein transferase activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline