This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FEZ2 SIRNA Assays ›
Gene Symbol:
Fez2
Gene Name:
fasciculation and elongation protein zeta 2 (zygin II)
Gene Aliases:
9030616F16, D17Ertd315e
Chromosome Location:
Chr.17: 78369212 - 78418152 on Build GRCm38
UniGene ID:
Mm.236835
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 171884

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fez2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001285940.1 NP_001272869.1 1, 2 326
NM_001285946.1 NP_001272875.1 1, 2 326
NM_001285949.1 NP_001272878.1 1, 2 326
NM_199448.3 NP_955519.1 1, 2 326
GenBank mRNA AK033547.1 - 1, 2 268
AK150241.1 - 1, 2 292
AK157221.1 - 1, 2 293
AK159545.1 - 1, 2 279
AK165595.1 - 1, 2 276
AY382580.1 - 1, 2 277
BC052366.1 - 1, 2 305
BC064130.1 - 1, 2 232
BC096619.1 - 1, 2 271 6391451

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001042548, NM_005102, BC110386, BC037566, BC018032, and others.
This siRNA also targets the following transcripts in Rat: NM_053600, XM_006239637, XM_006239639, XM_006239638, AB076183, and others.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mvc76:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline