This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PALMD SIRNA Assays ›
Gene Symbol:
Palmd
Gene Name:
palmdelphin
Gene Aliases:
4631423C22Rik, PALML
Chromosome Location:
Chr.3: 116918262 - 116968952 on Build GRCm38
UniGene ID:
Mm.253736
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 171937

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Palmd
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023245.3 NP_075734.3 8 2117
GenBank mRNA AF263246.1 - 8 1821
AJ312215.1 - 8 1840
AK014531.1 - 8 1915
AK171869.1 - 8 1848
AL391036.1 - 2 464
AL391037.1 - 8 1821
BC010193.1 - 8 1881 3156945

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of cell shape
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-584499df9d-64pmj:80/100.66.78.233:80.
git-commit: 256cec658627a60c8e1196038bf5905a13cd45ba
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0