This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LAPTM4B SIRNA Assays ›
Gene Symbol:
Laptm4b
Gene Name:
lysosomal-associated protein transmembrane 4B
Gene Aliases:
C330023P13Rik, LAPTM4beta
Chromosome Location:
Chr.15: 34238026 - 34284295 on Build GRCm38
UniGene ID:
Mm.197518
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 172190

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Laptm4b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033521.3 NP_277056.1 7 1025
GenBank mRNA AK077269.1 - 7 888
AK077933.1 - 7 1025
AK079581.1 - 7 888
AK082807.1 - 7 904
AK146868.1 - 6 791
AK166014.1 - 7 927
AK196114.1 - 1 35
AK200493.1 - 1 35
BC019120.1 - 7 895 4507607
EU561010.1 - 7 1025

Back To Top

More Information


Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0