This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other MTHFR SIRNA Assays ›
Gene Symbol:
MTHFR
Gene Name:
methylenetetrahydrofolate reductase (NAD(P)H)
Gene Aliases:
-
Chromosome Location:
Chr.1: 11785730 - 11806103 on Build GRCh38
UniGene ID:
Hs.214142
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 17299

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MTHFR
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005957.4 NP_005948.3 9, 10 1767
XM_005263458.3 XP_005263515.1 9, 10 2604
XM_005263460.4 XP_005263517.1 9, 10 2222
XM_005263462.4 XP_005263519.1 9, 10 1655
XM_005263463.3 XP_005263520.1 9, 10 1784
XM_011541495.2 XP_011539797.1 9, 10 2601
XM_011541496.2 XP_011539798.1 9, 10 2604
XM_017001328.1 XP_016856817.1 9, 10 2604
GenBank mRNA AB209113.1 - 8, 9 1805
AJ237672.1 - 8, 9 1667
AK312907.1 - 9, 10 1628
BC053509.1 - 9, 10 1589 6374885
JQ429447.1 - 5, 6 1043

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
cellular amino acid metabolic process
methionine metabolic process
blood circulation
regulation of histone methylation
response to vitamin B2
tetrahydrofolate interconversion
response to drug
response to amino acid
S-adenosylmethionine metabolic process
folic acid metabolic process
homocysteine metabolic process
response to folic acid
oxidation-reduction process
response to interleukin-1
heterochromatin maintenance
methylenetetrahydrofolate reductase (NAD(P)H) activity
protein complex binding
flavin adenine dinucleotide binding
NADP binding
modified amino acid binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline