This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other OBFC1 SIRNA Assays ›
Gene Symbol:
Obfc1
Gene Name:
oligonucleotide/oligosaccharide-binding fold containing 1
Gene Aliases:
0610009H20Rik, 2310057J23Rik, AAF-44, AAF44, AI413458, RPA-32
Chromosome Location:
Chr.19: 47501031 - 47545776 on Build GRCm38
UniGene ID:
Mm.218048
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 173740

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Obfc1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175360.2 NP_780569.1 5 641
GenBank mRNA AK075574.1 - 5 641
AK135228.1 - 5 659
AK146929.1 - 5 719
AK164386.1 - 5 651
BC029548.1 - 5 681 5012064

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

telomere maintenance
telomere maintenance via telomere lengthening
positive regulation of DNA replication
nucleic acid binding
DNA binding
single-stranded DNA binding
protein binding
single-stranded telomeric DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-5p4xw:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline