siRNA Details
| Gene Symbol: Desi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024282.3 | NP_077244.1 | 5 | 2442 | ||
| GenBank mRNA | AK020014.1 | - | 5 | 2327 | ||
| AK029574.1 | - | 6 | 2518 | |||
| AK146492.1 | - | 5 | 2175 | |||
| BC046816.1 | - | 5 | 2284 | 5702476 | ||

Desi2
Genomic Map