This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other WBSCR27 SIRNA Assays ›
Gene Symbol:
Wbscr27
Gene Name:
Williams Beuren syndrome chromosome region 27 (human)
Gene Aliases:
AW492986, BC002286
Chromosome Location:
Chr.5: 134932310 - 134942637 on Build GRCm38
UniGene ID:
Mm.34235
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 174377

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Wbscr27
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024479.2 NP_077799.2 2 233
GenBank mRNA AK042578.1 - 2 124
AK081683.1 - 2 252
AK132481.1 - 2 222
AY138964.1 - 2 233
AY354925.2 - 2 233
AY354926.1 - 2 134
AY354927.1 - 2 233
BC002286.1 - 2 224 3589835

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001109499, XM_006249191, XM_006249190.

Panther Classification:

Molecular Function -

methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
methylation
molecular_function
S-adenosylmethionine-dependent methyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline