This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other STPG1 SIRNA Assays ›
Gene Symbol:
Stpg1
Gene Name:
sperm tail PG rich repeat containing 1
Gene Aliases:
4930403G18Rik, 4930555I21Rik, Mapo2
Chromosome Location:
Chr.4: 135494833 - 135537803 on Build GRCm38
UniGene ID:
Mm.374844
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 174830

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Stpg1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030189.3 NP_084465.1 2 281
GenBank mRNA AK015061.1 - 3 476
AK019766.1 - 2 282

Back To Top

More Information


Panther Classification:

Molecular Function -

structural protein

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-875b6bdcc-jxtb8:80/100.66.75.28:80.
git-commit: 41fbaa8c9960493fb804edf63312caa25f4bdf16
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0