This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CWC22 SIRNA Assays ›
Gene Symbol:
Cwc22
Gene Name:
CWC22 spliceosome-associated protein homolog (S. cerevisiae)
Gene Aliases:
AA684037, AI173004, AL022752, B230213M24, mKIAA1604
Chromosome Location:
Chr.2: 77895653 - 77946356 on Build GRCm38
UniGene ID:
Mm.288151
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 174921

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cwc22
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290740.1 NP_001277669.1 4 480
NM_030560.5 NP_085037.2 5 538
NM_172667.2 NP_766255.1 4 477
GenBank mRNA AK045589.1 - 4 309
AK051237.1 - 4 305
AK077961.1 - 5 366
AK088752.1 - 4 308
AK132001.1 - 4 421
AK143352.1 - 4 310
AK149528.1 - 4 304
BC003993.1 - 4 290 3492146
BU613482.1 - 4 477

Back To Top

More Information


Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
RNA splicing
RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline