This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other BC004004 SIRNA Assays ›
Gene Symbol:
BC004004
Gene Name:
cDNA sequence BC004004
Gene Aliases:
2400006G15Rik
Chromosome Location:
Chr.17: 29268788 - 29302887 on Build GRCm38
UniGene ID:
Mm.290116
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 174924

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: BC004004
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030561.3 NP_085038.3 7 1039
GenBank mRNA AK030596.1 - 5 609
AK180627.1 - 3 174
AK180865.1 - 260
AK198839.1 - 3 174
BC004004.1 - 7 876 3492768
BC058575.1 - 7 848 6488611

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_199379, XM_006256174, XM_006256180, XM_008772724, XM_006256172, and others.

Gene Ontology Categories:

Function(s) Process(es)

wound healing
positive regulation of cell cycle
epithelial cell proliferation
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-5p4xw:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline