This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other HEMGN SIRNA Assays ›
Gene Symbol:
Hemgn
Gene Name:
hemogen
Gene Aliases:
4921524M03Rik, AI317176, EDAG, Hgn, mNDR
Chromosome Location:
Chr.4: 46393989 - 46413504 on Build GRCm38
UniGene ID:
Mm.25793
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 175519

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 286.00
Your Price
Online offer: 277.65
277.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hemgn
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053149.2 NP_444379.1 3 385
GenBank mRNA AF269248.1 - 3 385
AK006588.1 - 4 400
AK076587.1 - 4 370
AK077692.1 - 3 228
AK156536.1 - 3 313
AK171890.1 - 3 304
AK171906.1 - 3 434
AK172421.1 - 3 304
AY699986.1 - 3 227
BC050798.1 - 4 364 6743819

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_133294, XM_008763694, XM_008763693, BC078739, AJ302650.

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
cell differentiation
regulation of osteoblast differentiation
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline