This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TTYH3 SIRNA Assays ›
Gene Symbol:
Ttyh3
Gene Name:
tweety homolog 3 (Drosophila)
Gene Aliases:
2900029G13Rik, AI414930, TTY3, mTTY3
Chromosome Location:
Chr.5: 140620578 - 140649031 on Build GRCm38
UniGene ID:
Mm.28947
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 176290

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ttyh3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK052340.1 - 7 1033
AK173233.1 - 4 422
BC059083.1 - 7 1033 6827289
BC062917.1 - 7 1039 6831835

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107124, XM_008768969.

Panther Classification:

Molecular Function -

ion channel

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline