This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PSME1 SIRNA Assays ›
Gene Symbol:
PSME1
Gene Name:
proteasome activator subunit 1
Gene Aliases:
HEL-S-129m, IFI5111, PA28A, PA28alpha, REGalpha
Chromosome Location:
Chr.14: 24136158 - 24138967 on Build GRCh38
UniGene ID:
Hs.75348
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 17637

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PSME1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281528.1 NP_001268457.1 7 542
NM_001281529.1 NP_001268458.1 7 542
NM_006263.3 NP_006254.1 7 542
NM_176783.2 NP_788955.1 7 542
GenBank mRNA AK311490.1 - 5 743
AK312211.1 - 7 477
BC000352.2 - 7 528 2961448
BC007503.2 - 7 528 2961448
BM470561.1 - 7 478
BQ060125.1 - 7 500
BT019337.1 - 7 437
BU192084.1 - 7 473
BX248293.1 - 3 210
CR456780.1 - 7 437
CR541658.1 - 7 437
DQ185047.1 - 5 372
GQ472237.1 - 7 528
L07633.1 - 7 529

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
protein polyubiquitination
stimulatory C-type lectin receptor signaling pathway
antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
regulation of cellular amino acid metabolic process
positive regulation of endopeptidase activity
anaphase-promoting complex-dependent catabolic process
tumor necrosis factor-mediated signaling pathway
NIK/NF-kappaB signaling
Fc-epsilon receptor signaling pathway
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of mRNA stability
T cell receptor signaling pathway
negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
Wnt signaling pathway, planar cell polarity pathway
regulation of proteasomal protein catabolic process
negative regulation of canonical Wnt signaling pathway
positive regulation of canonical Wnt signaling pathway
regulation of G1/S transition of mitotic cell cycle
endopeptidase activator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-m9fgr:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline