This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PDGFD SIRNA Assays ›
Gene Symbol:
Pdgfd
Gene Name:
platelet-derived growth factor, D polypeptide
Gene Aliases:
1110003I09Rik
Chromosome Location:
Chr.9: 6168362 - 6536014 on Build GRCm38
UniGene ID:
Mm.390122
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 177236

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pdgfd
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027924.2 NP_082200.1 7 1341
GenBank mRNA AK003359.2 - 7 1343
AK141551.1 - 7 1352
BC030896.1 - 7 1077 4489485

Back To Top

More Information


Panther Classification:

Molecular Function -

growth factor

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
cellular process
platelet-derived growth factor receptor signaling pathway
positive regulation of fibroblast proliferation
positive regulation of smooth muscle cell proliferation
regulation of peptidyl-tyrosine phosphorylation
positive regulation of cell division
cellular response to amino acid stimulus
cellular response to transforming growth factor beta stimulus
positive regulation of smooth muscle cell chemotaxis
positive regulation of glomerular mesangial cell proliferation
positive regulation of monocyte extravasation
platelet-derived growth factor receptor binding
growth factor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-hssmq:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline