This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other KIF1BP SIRNA Assays ›
Gene Symbol:
Kif1bp
Gene Name:
KIF1 binding protein
Gene Aliases:
0710007C18Rik, 2510003E04Rik, KBP, mKIAA1279
Chromosome Location:
Chr.10: 62558470 - 62578464 on Build GRCm38
UniGene ID:
Mm.258955
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 177515

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kif1bp
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028197.2 NP_082473.2 7 2091
GenBank mRNA AK010898.1 - 1 344
AK129320.1 - 7 2076
AK141324.1 - 7 2100
AK149629.1 - 7 2029
AK158730.1 - 8 1825
AK198768.1 - 1 136
AK201307.1 - 1 135
BC038828.1 - 3 1400
BC046485.1 - 7 2062 5350638
BC055307.1 - 7 2045 6493547

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitochondrial transport
multicellular organism development
nervous system development
cell differentiation
kinesin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-rp79v:80/100.66.77.211:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline