siRNA Details
| Gene Symbol: 1700034I23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006596.1 | - | 1 | 1131 | ||
| AK077003.1 | - | 1 | 1089 | |||
| BC117739.1 | - | 1 | 1097 | 40090015 | ||
| Gene Symbol: Gm10731 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK077188.1 | - | 1 | 1082 | ||

1700034I23Rik
Genomic Map