siRNA Details
| Gene Symbol: ZNHIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006349.2 | NP_006340.1 | 3, 4 | 767 | ||
| XM_011515739.2 | XP_011514041.1 | 3, 4 | 498 | |||
| GenBank mRNA | AF093571.1 | - | 3, 4 | 345 | ||
| AK125123.1 | - | 4, 5 | 3764 | |||
| BC017333.2 | - | 3, 4 | 767 | 4647446 | ||
| CR456992.1 | - | 3, 4 | 275 | |||
| U61837.1 | - | 3, 4 | 305 | |||

ZNHIT1
Genomic Map