siRNA Details
| Gene Symbol: Taf7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028958.4 | NP_083234.2 | 6, 7 | 769 | ||
| GenBank mRNA | AF285574.1 | - | 6, 7 | 573 | ||
| AK017109.1 | - | 6, 7 | 770 | |||
| BC106854.2 | - | 6, 7 | 561 | 40046501 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Taf7l| Gene Symbol: Taf7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028958.4 | NP_083234.2 | 6, 7 | 769 | ||
| GenBank mRNA | AF285574.1 | - | 6, 7 | 573 | ||
| AK017109.1 | - | 6, 7 | 770 | |||
| BC106854.2 | - | 6, 7 | 561 | 40046501 | ||