This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SSMEM1 SIRNA Assays ›
Gene Symbol:
Ssmem1
Gene Name:
serine-rich single-pass membrane protein 1
Gene Aliases:
1700016K02Rik, 1700025E21Rik
Chromosome Location:
Chr.6: 30509849 - 30520253 on Build GRCm38
UniGene ID:
Mm.46117
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 178351

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 274.65
274.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ssmem1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027073.1 NP_081349.1 2 486
NM_029373.4 NP_083649.1 2 300
GenBank mRNA AK006038.1 - 2 487
AK006332.1 - 2 286
BC049755.1 - 2 435 6773255
BU938696.1 - 2 302

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-r9tfj:80/100.66.76.159:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline